Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118104.5 | NP_193719.1 | 1 - 2 | 772 | 66 | |
GenBank mRNA | AY090355.1 | - | 1 - 2 | 300 | 66 | |
AY122906.1 | - | 1 - 2 | 240 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |