Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341398.1 | NP_001329687.1 | 10 - 11 | 1420 | 64 | |
NM_118126.4 | NP_193740.1 | 10 - 11 | 1464 | 64 | ||
GenBank mRNA | BT025334.1 | - | 10 - 11 | 1298 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |