Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341400.1 | NP_001327970.1 | 3 - 4 | 477 | 153 | |
NM_001341401.1 | NP_001327969.1 | 1 - 2 | 157 | 153 | ||
NM_118132.5 | NP_193746.3 | 4 - 5 | 570 | 153 | ||
GenBank mRNA | AK230206.1 | - | 4 - 5 | 492 | 153 | |
BT004184.1 | - | 4 - 5 | 484 | 153 | ||
BT005475.1 | - | 4 - 5 | 473 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |