Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203853.2 | NP_001190782.1 | 7 - 8 | 2254 | 150 | |
NM_118209.3 | NP_567616.1 | 6 - 7 | 2399 | 150 | ||
GenBank mRNA | AF327068.1 | - | 6 - 7 | 2181 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |