Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118218.1 | NP_193832.1 | 4 - 5 | 514 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |