Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203855.2 | NP_001190784.2 | 4 - 5 | 491 | 64 | |
NM_001203856.1 | NP_001190785.1 | 5 - 6 | 642 | 64 | ||
NM_118234.4 | NP_193847.2 | 5 - 6 | 603 | 64 | ||
GenBank mRNA | AY058845.1 | - | 5 - 6 | 603 | 64 | |
AY079034.1 | - | 4 - 5 | 491 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |