Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118246.2 | NP_193859.1 | 12 - 13 | 1975 | 110 | |
GenBank mRNA | D11371.1 | - | 12 - 13 | 1942 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |