Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118246.2 | NP_193859.1 | 9 - 10 | 1559 | 74 | |
GenBank mRNA | D11371.1 | - | 9 - 10 | 1526 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |