Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341491.1 | NP_001329575.1 | 6 - 7 | 1310 | 135 | |
NM_118269.6 | NP_193880.5 | 6 - 7 | 1236 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |