Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084951.2 | NP_001078420.1 | 3 - 4 | 395 | 69 | |
NM_118279.3 | NP_567630.1 | 2 - 3 | 496 | 69 | ||
GenBank mRNA | AY085756.1 | - | 2 - 3 | 451 | 69 | |
BT026061.1 | - | 2 - 3 | 293 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |