Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341506.1 | NP_001329633.1 | 1 - 2 | 166 | 73 | |
NM_118281.4 | NP_193892.1 | 1 - 2 | 259 | 73 | ||
GenBank mRNA | BT011760.1 | - | 1 - 2 | 259 | 73 | |
BT012384.1 | - | 1 - 2 | 166 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |