Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118287.5 | NP_193898.3 | 13 - 14 | 3111 | 75 | |
GenBank mRNA | AY557186.1 | - | 13 - 14 | 2589 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |