Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125558.1 | NP_001119030.1 | 6 - 7 | 909 | 116 | |
NM_118334.5 | NP_193944.2 | 10 - 11 | 1602 | 116 | ||
GenBank mRNA | AY518293.1 | - | 10 - 11 | 1365 | 116 | |
FJ708751.1 | - | 10 - 11 | 1257 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |