Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341553.1 | NP_001328218.1 | 10 - 11 | 2335 | 77 | |
NM_118380.3 | NP_567662.1 | 10 - 11 | 2558 | 77 | ||
NM_202863.3 | NP_974592.1 | 10 - 11 | 1924 | 77 | ||
GenBank mRNA | AY052361.1 | - | 10 - 11 | 2443 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |