Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341552.1 | NP_001328217.1 | 1 - 2 | 723 | 116 | |
NM_118380.3 | NP_567662.1 | 1 - 2 | 757 | 116 | ||
GenBank mRNA | AY052361.1 | - | 1 - 2 | 642 | 116 | |
BT001093.1 | - | 1 - 2 | 390 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |