Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118393.3 | NP_567663.1 | 7 - 8 | 1060 | 90 | |
GenBank mRNA | AY059803.1 | - | 7 - 8 | 967 | 90 | |
AY087472.1 | - | 7 - 8 | 954 | 90 | ||
BT003349.1 | - | 7 - 8 | 811 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |