Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR5-LIKE A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084962.1 | NP_001078431.1 | 4 - 5 | 675 | 104 | |
NM_001084963.1 | NP_001078432.1 | 4 - 5 | 678 | 104 | ||
NM_001341564.1 | NP_001320036.1 | 4 - 5 | 630 | 104 | ||
NM_118418.4 | NP_567672.1 | 4 - 5 | 684 | 104 | ||
NM_179092.1 | NP_849423.1 | 4 - 5 | 684 | 104 | ||
GenBank mRNA | BT000905.1 | - | 4 - 5 | 513 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |