Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118420.5 | NP_194022.3 | 5 - 6 | 1502 | 117 | |
GenBank mRNA | AY099585.1 | - | 5 - 6 | 1233 | 117 | |
AY128834.1 | - | 5 - 6 | 1167 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |