Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341565.1 | NP_001328989.1 | 4 - 5 | 1079 | 99 | |
NM_118421.4 | NP_567673.1 | 3 - 4 | 1000 | 99 | ||
GenBank mRNA | AY063900.1 | - | 3 - 4 | 626 | 99 | |
AY096504.1 | - | 3 - 4 | 477 | 99 | ||
AY850161.1 | - | 3 - 4 | 785 | 99 | ||
DQ437531.1 | - | 3 - 4 | 785 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |