Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341579.1 | NP_001328631.1 | 3 - 4 | 1122 | 133 | |
NM_001341580.1 | NP_001328630.1 | 3 - 4 | 1122 | 133 | ||
NM_118447.3 | NP_567679.2 | 3 - 4 | 1264 | 133 | ||
GenBank mRNA | AK117765.1 | - | 3 - 4 | 1122 | 133 | |
BT005954.1 | - | 3 - 4 | 1088 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |