Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036628.2 | NP_001031705.2 | 1 - 2 | 295 | 95 | |
NM_118491.3 | NP_194091.1 | 1 - 2 | 535 | 95 | ||
GenBank mRNA | AY099811.1 | - | 1 - 2 | 295 | 95 | |
BT000307.1 | - | 1 - 2 | 257 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |