Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126396.2 | NP_178444.1 | 1 - 2 | 829 | 116 | |
GenBank mRNA | AF492661.1 | - | 1 - 2 | 689 | 116 | |
AK228718.1 | - | 1 - 2 | 830 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |