Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126402.3 | NP_178450.2 | 5 - 6 | 676 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |