Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335192.1 | NP_001324930.1 | 5 - 6 | 854 | 97 | |
NM_126409.4 | NP_565303.1 | 4 - 5 | 1069 | 97 | ||
GenBank mRNA | AY085962.1 | - | 4 - 5 | 841 | 97 | |
BT029190.1 | - | 4 - 5 | 642 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |