Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335203.1 | NP_001325274.1 | 8 - 9 | 1057 | 96 | |
NM_126426.5 | NP_178474.2 | 8 - 9 | 993 | 96 | ||
GenBank mRNA | AB091252.1 | - | 8 - 9 | 892 | 96 | |
AK228782.1 | - | 8 - 9 | 894 | 96 | ||
BT022079.1 | - | 8 - 9 | 880 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |