Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341000.1 | NP_001329358.1 | 9 - 10 | 996 | 85 | |
NM_001341006.1 | NP_001329362.1 | 11 - 12 | 1323 | 85 | ||
NM_001341007.1 | NP_001329361.1 | 10 - 11 | 1433 | 85 | ||
NM_001341008.1 | NP_001329366.1 | 10 - 11 | 1366 | 85 | ||
NM_148326.2 | NP_680692.1 | 8 - 9 | 958 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |