Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341092.1 | NP_001319956.1 | 18 - 19 | 2545 | 88 | |
NM_117725.3 | NP_193363.4 | 18 - 19 | 1975 | 88 | ||
NM_148347.2 | NP_680713.2 | 19 - 20 | 2341 | 88 | ||
NM_179211.3 | NP_849542.2 | 17 - 18 | 4776 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |