Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341092.1 | NP_001319956.1 | 5 - 6 | 1034 | 93 | |
NM_117725.3 | NP_193363.4 | 5 - 6 | 621 | 93 | ||
NM_148347.2 | NP_680713.2 | 6 - 7 | 830 | 93 | ||
NM_179211.3 | NP_849542.2 | 4 - 5 | 3265 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |