Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148462.3 | NP_683303.1 | 1 | 590 | 143 | |
GenBank mRNA | AK221719.1 | - | 1 | 451 | 143 | |
AY085299.1 | - | 1 | 452 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |