Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333839.1 | NP_001319269.1 | 6 - 7 | 1217 | 91 | |
NM_001333840.1 | NP_001319268.1 | 6 - 7 | 1256 | 91 | ||
NM_001333841.1 | NP_001322290.1 | 6 - 7 | 1219 | 91 | ||
NM_001333842.1 | NP_001322291.1 | 5 - 6 | 1326 | 91 | ||
NM_001333843.1 | NP_001322289.1 | 6 - 7 | 1137 | 91 | ||
NM_148599.4 | NP_683440.2 | 5 - 6 | 943 | 91 | ||
GenBank mRNA | KJ920364.1 | - | 5 - 6 | 893 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |