Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332873.1 | NP_001322311.1 | 14 - 15 | 1332 | 71 | |
NM_102715.8 | NP_174268.7 | 14 - 15 | 1453 | 71 | ||
NM_180624.6 | NP_850955.5 | 13 - 14 | 1565 | 71 | ||
GenBank mRNA | AY091006.1 | - | 13 - 14 | 1542 | 71 | |
AY142598.1 | - | 13 - 14 | 1103 | 71 | ||
FJ708642.1 | - | 13 - 14 | 1103 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |