Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332873.1 | NP_001322311.1 | 16 - 17 | 1419 | 69 | |
NM_102715.8 | NP_174268.7 | 16 - 17 | 1540 | 69 | ||
NM_180624.6 | NP_850955.5 | 15 - 16 | 1652 | 69 | ||
GenBank mRNA | AY091006.1 | - | 15 - 16 | 1629 | 69 | |
AY142598.1 | - | 15 - 16 | 1190 | 69 | ||
FJ708642.1 | - | 15 - 16 | 1190 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |