Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332873.1 | NP_001322311.1 | 17 - 18 | 1816 | 81 | |
NM_102715.8 | NP_174268.7 | 17 - 18 | 1937 | 81 | ||
NM_180624.6 | NP_850955.5 | 16 - 17 | 2049 | 81 | ||
GenBank mRNA | AY091006.1 | - | 16 - 17 | 2026 | 81 | |
AY142598.1 | - | 16 - 17 | 1587 | 81 | ||
FJ708642.1 | - | 16 - 17 | 1587 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |