Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036039.2 | NP_001031116.1 | 22 - 23 | 3707 | 78 | |
NM_102777.3 | NP_174329.1 | 23 - 24 | 3645 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |