Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036039.2 | NP_001031116.1 | 2 - 3 | 989 | 89 | |
NM_102777.3 | NP_174329.1 | 3 - 4 | 927 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |