Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332901.1 | NP_001319113.1 | 4 - 5 | 1087 | 122 | |
NM_102782.4 | NP_174334.2 | 4 - 5 | 1095 | 122 | ||
GenBank mRNA | EU250988.1 | - | 4 - 5 | 1069 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |