Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102785.5 | NP_174337.1 | 6 - 7 | 1143 | 71 | |
GenBank mRNA | AJ440967.1 | - | 6 - 7 | 794 | 71 | |
AK226570.1 | - | 6 - 7 | 1008 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |