Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332903.1 | NP_001322322.1 | 5 - 6 | 1277 | 98 | |
NM_001332904.1 | NP_001322320.1 | 5 - 6 | 797 | 98 | ||
NM_001332905.1 | NP_001322321.1 | 5 - 6 | 797 | 98 | ||
NM_102789.5 | NP_174340.2 | 5 - 6 | 1277 | 98 | ||
GenBank mRNA | AY070739.1 | - | 5 - 6 | 690 | 98 | |
AY250843.1 | - | 5 - 6 | 635 | 98 | ||
BT010462.1 | - | 5 - 6 | 635 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |