Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084165.1 | NP_001077634.1 | 2 - 3 | 442 | 82 | |
NM_001332933.1 | NP_001322001.1 | 2 - 3 | 482 | 82 | ||
NM_102836.3 | NP_564369.1 | 2 - 3 | 460 | 82 | ||
GenBank mRNA | AF360277.1 | - | 2 - 3 | 458 | 82 | |
AY051039.1 | - | 2 - 3 | 302 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |