Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084170.2 | NP_001077639.1 | 1 - 2 | 164 | 135 | |
NM_001332956.1 | NP_001319121.1 | 1 - 2 | 162 | 135 | ||
NM_001332957.1 | NP_001322841.1 | 1 - 2 | 162 | 135 | ||
NM_001332958.1 | NP_001322840.1 | 1 - 2 | 162 | 135 | ||
NM_001332959.1 | NP_001322843.1 | 1 - 2 | 162 | 135 | ||
NM_001332960.1 | NP_001322842.1 | 1 - 2 | 162 | 135 | ||
NM_001332961.1 | NP_001322844.1 | 1 - 2 | 161 | 135 | ||
NM_102875.4 | NP_174421.3 | 1 - 2 | 166 | 135 | ||
GenBank mRNA | AJ404471.1 | - | 1 - 2 | 151 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |