Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198193.1 | NP_001185122.1 | 2 - 3 | 443 | 128 | |
NM_001332962.1 | NP_001320669.1 | 1 - 2 | 840 | 128 | ||
NM_102881.4 | NP_174427.3 | 2 - 3 | 425 | 128 | ||
GenBank mRNA | FJ708643.1 | - | 1 - 2 | 98 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |