Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102886.2 | NP_174432.2 | 1 - 2 | 434 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |