Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102933.3 | NP_174479.1 | 10 - 11 | 1418 | 81 | |
GenBank mRNA | AY080680.1 | - | 10 - 11 | 1314 | 81 | |
AY117298.1 | - | 10 - 11 | 1260 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |