Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102940.6 | NP_174486.1 | 3 - 4 | 1168 | 53 | |
GenBank mRNA | AY044335.1 | - | 3 - 4 | 699 | 53 | |
AY084576.1 | - | 3 - 4 | 808 | 53 | ||
AY128355.1 | - | 3 - 4 | 805 | 53 | ||
BT000019.1 | - | 3 - 4 | 699 | 53 | ||
X58149.1 | - | 777 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |