Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102953.4 | NP_174499.1 | 9 - 10 | 1185 | 66 | |
NM_179407.3 | NP_849738.1 | 9 - 10 | 1185 | 66 | ||
GenBank mRNA | AY093169.1 | - | 9 - 10 | 1070 | 66 | |
BT008758.1 | - | 9 - 10 | 1012 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |