Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATDAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102954.4 | NP_174500.1 | 3 - 4 | 434 | 111 | |
GenBank mRNA | BT024553.1 | - | 3 - 4 | 220 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |