Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333001.1 | NP_001322280.1 | 3 - 4 | 1225 | 98 | |
NM_001333002.1 | NP_001322279.1 | 2 - 3 | 149 | 98 | ||
NM_102957.4 | NP_564392.1 | 3 - 4 | 1225 | 98 | ||
GenBank mRNA | AB493491.1 | - | 3 - 4 | 835 | 98 | |
AY048689.1 | - | 3 - 4 | 853 | 98 | ||
BT029211.1 | - | 3 - 4 | 835 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |