Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333007.1 | NP_001322345.1 | 6 - 7 | 1734 | 72 | |
NM_102972.3 | NP_174516.1 | 7 - 8 | 1322 | 72 | ||
GenBank mRNA | AF439851.1 | - | 7 - 8 | 1304 | 72 | |
AY125555.1 | - | 7 - 8 | 1023 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |