Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036050.2 | NP_001031127.1 | 4 - 5 | 1115 | 119 | |
NM_102974.5 | NP_564399.1 | 5 - 6 | 829 | 119 | ||
NM_202223.3 | NP_973952.1 | 5 - 6 | 897 | 119 | ||
GenBank mRNA | AB085684.1 | - | 5 - 6 | 683 | 119 | |
AF334723.1 | - | 4 - 5 | 523 | 119 | ||
AF385687.1 | - | 5 - 6 | 739 | 119 | ||
AY088560.1 | - | 5 - 6 | 721 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |