Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204026.2 | NP_001190955.1 | 10 - 11 | 901 | 97 | |
NM_001342505.1 | NP_001328584.1 | 9 - 10 | 1120 | 97 | ||
NM_001342506.1 | NP_001328583.1 | 10 - 11 | 967 | 97 | ||
NM_120018.4 | NP_195569.1 | 10 - 11 | 923 | 97 | ||
GenBank mRNA | AY086801.1 | - | 10 - 11 | 823 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |